Skip to Content
Merck
All Photos(1)

Key Documents

51788

Supelco

Aniline

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$39.40
5 ML
$117.00

$39.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$39.40
5 ML
$117.00

About This Item

Linear Formula:
C6H5NH2
CAS Number:
Molecular Weight:
93.13
Beilstein:
605631
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$39.40


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

3.22 (185 °C, vs air)

vapor pressure

0.7 mmHg ( 25 °C)

Assay

≥99.5% (GC)

autoignition temp.

1139 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

11 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.1% water
<0.03% nitrobenzene (C6H5NO2)

refractive index

n20/D 1.586 (lit.)
n20/D 1.586

bp

184 °C (lit.)

mp

−6 °C (lit.)

density

1.022 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Nc1ccccc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2

InChI key

PAYRUJLWNCNPSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl heneicosanoate has been used as an internal standard in the quantitative determination of fatty acids and their isomers composition using gas chromatography method of analysis (GC).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

The acute toxicity of aniline involves its activation in vivo to 4-hydroxyaniline and the formation of adducts with hemoglobin. In erythrocytes, this is associated with the release of iron and the accumulation of methemoglobin and the development of hemolytic anemia and inflammation of the spleen. Tumor formation is often observed in the spleen on prolonged administration.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teaching Innovations in Lipid Science (2007)
Conor J Kearney et al.
Science advances, 7(8) (2021-02-21)
Multimodal single-cell RNA sequencing enables the precise mapping of transcriptional and phenotypic features of cellular differentiation states but does not allow for simultaneous integration of critical posttranslational modification data. Here, we describe SUrface-protein Glycan And RNA-seq (SUGAR-seq), a method that
M Peluso et al.
Carcinogenesis, 21(2), 183-187 (2000-02-05)
The 'Mediterranean diet', a diet rich in cereals, fruit and vegetables, has been associated with lowering the risk of a variety of cancers of the digestive tract and the bladder. In a previous study, we showed that the high phenolic
Conghui Tang et al.
Journal of the American Chemical Society, 134(46), 18924-18927 (2012-11-08)
A novel and efficient copper-catalyzed azidation reaction of anilines via C-H activation has been developed. This method, in which the primary amine acts as a directing group by coordinating to the metal center, provides ortho azidation products regioselectively under mild
S A Vaziri et al.
Pharmacogenetics, 11(1), 7-20 (2001-02-24)
Arylamines such as 2-naphthylamine and 4-aminobiphenyl are suspected human bladder procarcinogens that require bioactivation to DNA-reactive species to exert their carcinogenic potential. The goals of the present study were (i) to assay for the presence of the arylamine acetyltransferases NAT1

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service