Skip to Content
Merck
All Photos(1)

Key Documents

78471

Sigma-Aldrich

Shellac wax-free

tested according to Ph. Eur.

Synonym(s):

Lacca

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jilai Gong et al.
Journal of environmental sciences (China), 24(7), 1165-1173 (2012-01-01)
This study describes a new effective adsorbent for cadmium removal from aqueous solution synthesized by coating a shellac layer, a natural biodegradable and renewable resin with abundant hydroxyl and carboxylic groups, on the surface of iron oxide magnetic nanoparticles. Transmission
A rare eyelid dermatitis allergen: shellac in a popular mascara.
Tatyana Shaw et al.
Dermatitis : contact, atopic, occupational, drug, 20(6), 341-345 (2009-12-05)
Shinjita Das et al.
Dermatitis : contact, atopic, occupational, drug, 22(4), 220-222 (2011-07-26)
Shellac is a common ingredient in cosmetics such as mascaras, lipsticks, nail polish, and hairsprays. As an emerging contact allergen in patients with eyelid dermatitis and contact cheilitis, it was added to the 2009-2010 North American Contact Dermatitis Group screening
Stefan Henning et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 221, 11-18 (2012-06-30)
Microscopic capsules made from polysaccharides are used as carriers for drugs and food additives. Here, we use NMR microscopy to assess the permeability of capsule membranes and their stability under different environmental conditions. The results allow us to determine the
Shwan A Hamad et al.
Physical chemistry chemical physics : PCCP, 15(7), 2337-2344 (2013-01-09)
We have developed a theoretical model for the kinetics of release of living cells from composite shellac-cell microcapsules. The model describes the kinetics of cell release from the microcapsules triggered by: (i) pH change, which dissolves the shellac and (ii)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service