Skip to Content
Merck
All Photos(1)

Key Documents

8.14732

Sigma-Aldrich

Diisodecyl phthalate

for synthesis

Synonym(s):

Diisodecyl phthalate, 1,2-Benzenedicarboxylic acid diisodecyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$63.30
1 L
$304.00

$63.30


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$63.30
1 L
$304.00

About This Item

Empirical Formula (Hill Notation):
C28H46O4
CAS Number:
Molecular Weight:
446.66
MDL number:
UNSPSC Code:
12352108
EC Index Number:
247-977-1
NACRES:
NA.22

$63.30


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

liquid

autoignition temp.

380 °C

potency

>5000 mg/kg LD50, oral (Rat)
>3160 mg/kg LD50, skin (Rabbit)

bp

250-267 °C/7 hPa

mp

-50 °C

transition temp

flash point 240 °C

solubility

0.00028 g/L

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCCCCCCC(C)C)C(=O)c1c(cccc1)C(=O)OCCCCCCCC(C)C

InChI

1S/C28H46O4/c1-23(2)17-11-7-5-9-15-21-31-27(29)25-19-13-14-20-26(25)28(30)32-22-16-10-6-8-12-18-24(3)4/h13-14,19-20,23-24H,5-12,15-18,21-22H2,1-4H3

InChI key

ZVFDTKUVRCTHQE-UHFFFAOYSA-N

Application

  • Evaluating phthalate contaminant migration using thermal desorption–gas chromatography–mass spectrometry (TD–GC–MS): This study investigates the migration of phthalates, including diisodecyl phthalate, from materials like polyvinyl chloride, providing insights crucial for assessing exposure risks associated with everyday materials (Ouchi et al., 2019).
  • Toxicity and estrogenic endocrine disrupting activity of phthalates and their mixtures: This research focuses on the health effects of various phthalates, emphasizing the potential endocrine-disrupting properties of compounds like diisodecyl phthalate, which are widely used in consumer products (Chen et al., 2014).

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Density (d 20 °C/ 4 °C): 0.963 - 0.968
Identity (IR): passes test

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

527.0 °F - closed cup

Flash Point(C)

275.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hello, we would like to use this product as a control measurement for the fogging test according to PV 3015. Please provide a reference value for this product for the fogging test according to PV 3015 ? Thank you.

    1 answer
    1. Unfortunately, this product is not tested against method PV 3015, so there is no information or data that can be provided.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service