Skip to Content
Merck
All Photos(1)

Key Documents

75741

Supelco

Ginkgolic acid C15:1

analytical standard

Synonym(s):

22:1-ω7-Anacardic acid, 6-[(8Z)-Pentadecenyl]-salicylic acid, Anacardic acid monoene, Ginkgolic acid I, Romanicardic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$532.00

$532.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$532.00

About This Item

Empirical Formula (Hill Notation):
C22H34O3
CAS Number:
Molecular Weight:
346.50
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$532.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥80% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CCCCCC\C=C/CCCCCCCc1cccc(O)c1C(O)=O

InChI

1S/C22H34O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-17-15-18-20(23)21(19)22(24)25/h7-8,15,17-18,23H,2-6,9-14,16H2,1H3,(H,24,25)/b8-7-

InChI key

YXHVCZZLWZYHSA-FPLPWBNLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginkgolic acid C15:1 is an alkylphenol belonging to the class of ginkgolic acids isolated from Ginkgo biloba leaves. It also shows strong allergenic properties, with carcinogenic and mutagenic activity.[1]

Application

Ginkgolic acid C15:1 may be used as a reference standard in the quantitative determination of alkylphenols like ginkgolic acids in Ginkgolic extract using high performance liquid chromatography coupled with mass spectrometry(HPLC-MS).
Ginkgolic acid C15:1 may be used as an analytical reference standard for the quantification of the analyte in Ginkgo biloba plant extracts, and dietary supplements using chromatography techniques.[2][3]
Ginkgolic acid C15:1 may be used as a reference standard in the quantitative determination of alkylphenols like ginkgolic acids in Ginkgolic extract using high performance liquid chromatography coupled with mass spectrometry(HPLC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

509.9 °F - (calculated)

Flash Point(C)

265.5 °C - (calculated)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple HPLC-UV method for the assay of ginkgolic acids in Ginkgo biloba extracts
Fuzzati.N, et al.
Fitoterapia, 74(3), 247-256 (2003)
Concurrent supercritical fluid chromatographic analysis of terpene lactones and ginkgolic acids in Ginkgo biloba extracts and dietary supplements
Wang M, et al.
Analytical and Bioanalytical Chemistry, 408(17), 4649-4660 (2016)
P Loungratana et al.
The American journal of Chinese medicine, 32(1), 33-48 (2004-05-25)
A competitive enzyme-linked immunosorbent assay (ELISA) for ginkgolic acids (GAs) was developed using monoclonal antibody (MAb) 9F raised against 6-(13-formylheptyl) salicylic acid covalently coupled to bovine serum albumin (BSA). ELISA, at an effective measuring range of 300 ng/ml-1 microgram/ml of
Z H Liu et al.
Rapid communications in mass spectrometry : RCM, 23(13), 1899-1906 (2009-05-23)
Ginkgolic acids have been shown to possess allergenic as well as genotoxic and cytotoxic properties. The question arises whether the metabolism of ginkgolic acids in the liver could decrease or increase their toxicity. In this study, the in vitro metabolism
Joonseok Oh et al.
Journal of enzyme inhibition and medicinal chemistry, 28(3), 565-568 (2012-03-03)
Fatty acid synthase (FAS) has been proposed to be a new drug target for the development of anticancer agents because of the significant difference in expression of FAS between normal and tumour cells. Since a n-hexane-soluble extract from Ginkgo biloba

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service