Skip to Content
Merck
Ginkgolic acid C15:1 primary reference standard

Ginkgolic acid C15:1

Ginkgolic acid C15:1 primary reference standard
Synonym(s):
22:1-ω7-Anacardic acid, 6-[(8Z)-Pentadecenyl]-salicylic acid, Anacardic acid monoene, Ginkgolic acid I, Romanicardic acid
Empirical Formula (Hill Notation):
C22H34O3
CAS Number:
Molecular Weight:
346.50

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3