Skip to Content
Merck
All Photos(2)

Key Documents

73709

Supelco

(±)-γ-Hexabromocyclododecane

analytical standard

Synonym(s):

γ-HBCD, (1RS,2RS,5RS,6SR,9SR,10RS)-1,2,5,6,9,10-Hexabromocyclododecane

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$531.00

$531.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$531.00

About This Item

Empirical Formula (Hill Notation):
C12H18Br6
CAS Number:
Molecular Weight:
641.70
UNSPSC Code:
77101502
NACRES:
NA.24

$531.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

mp

208-214 °C

format

neat

SMILES string

Br[C@H]1CC[C@H](Br)[C@H](Br)CC[C@@H](Br)[C@H](Br)CC[C@H]1Br

InChI key

DEIGXXQKDWULML-MOCCIAMBSA-N

Related Categories

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service