Skip to Content
Merck
All Photos(1)

Key Documents

07444

Supelco

2,3,4,5,6-Pentabromotoluene

analytical standard

Synonym(s):

Pentabromotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$140.00

$140.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$140.00

About This Item

Linear Formula:
C6Br5CH3
CAS Number:
Molecular Weight:
486.62
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$140.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)
98.0-102.0% (argentometric after oxygen combustion)

shelf life

limited shelf life, expiry date on the label

mp

285-286 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

Cc1c(Br)c(Br)c(Br)c(Br)c1Br

InChI

1S/C7H3Br5/c1-2-3(8)5(10)7(12)6(11)4(2)9/h1H3

InChI key

OZHJEQVYCBTHJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2,3,4,5,6-Pentabromotoluene is a brominated compound, widely used as a flame retardant in textiles, rubber, unsaturated polyesters, polyethylene, SBR latex, etc.[1][2]

Application

2,3,4,5,6-Pentabromotoluene may be used as an analytical reference standard for the determination of the analyte in water[1] and soil samples[3] using gas chromatography technique.[1][3]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adelie penguin colonies as indicators of brominated flame retardants (BFRs) in East Antarctica
Lewis JP, et al.
Chemosphere (2020)
Gardner's Commercially Important Chemicals: Synonyms, Trade Names, and Properties (2005)
Qihang Wu et al.
The Science of the total environment, 599-600, 181-187 (2017-05-06)
While the production of polybrominated diphenyl ethers (PBDEs) was gradually phased out in the last decade, they may still pose hidden danger to the environment due to their toxicity and persistence. On the other hand, alternative halogenated flame retardants (AHFRs)
P López et al.
Journal of chromatography. A, 1216(3), 334-345 (2008-09-03)
Brominated flame retardants (BFRs) are the chemicals of high importance within the REAch framework. In addition to polybrominated diphenyl ethers (PBDEs), hexabromocyclododecane (HBCD) and tetrabromobisphenol A (TBBPA), other BFRs such as bromophenols, intermediates in FR formulation like bromoanilines, and their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service