Skip to Content
Merck
All Photos(2)

Key Documents

71008ATMBC

Millipore

Assurance® GDS Salmonella Tq Kit for Automation

suitable for PCR testing in Food and Beverages applications

Synonym(s):

Nucleic acid amplification system

Sign Into View Organizational & Contract Pricing

Select a Size

576 TESTS
$10,550.00

$10,550.00


Check Cart for Availability

Request more information

Select a Size

Change View
576 TESTS
$10,550.00

About This Item

UNSPSC Code:
41106302
NACRES:
NA.85

$10,550.00


Check Cart for Availability

Request more information

Product Name

GDS Salmonella Tq Automation Kit, BioControl, Molecular based PCR test for the detection of Salmonella spp. in food and environmental samples

Agency

PCR, Immunomagnetic Separation (IMS)
AFNOR
AOAC® (OMA)
AOAC® (PTM)
Health Canada
MicroVal

Quality Level

product line

Assurance GDS®

shelf life

6‑12 mo.

packaging

pkg of 576 tests

manufacturer/tradename

BioControl Systems 71008-576ATM

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl GDS
for use with GDS Rotor-Gene

storage temp.

2-8°C

suitability

Salmonella spp.

General description

GDS Salmonella Tq Automation Kit is an automated nucleic acid amplification system for the detection of Salmonella in food and environmental samples. The proprietary PickPen® IMS sample preparation procedure quickly and effectively captures Salmonella, separating the pathogen from inhibitory background materials and ensuring valid PCR reactions for all sample types, including challenging food and environmental samples. Assurance® GDS PCR technology provides two additional levels of specificity using both highly specific primers and probes to ensure accurate detection of Salmonella.

Application

GDS for Salmonella Tq is an automated nucleic acid amplification system for the detection of Salmonella in foods, ingredients, and environmental samples.

Features and Benefits

  • Next day results for foods utilizing a single enrichment media
  • Harmonized enrichment medias for testing samples for other pathogen targets in addition to Salmonella.
  • Validated for raw beef, raw pork, raw ground turkey, poultry carcass rinse, raw shrimp, non fat dry milk, liquid milk, egg, spices, peanut butter, pasta, stainless steel, rubber, concrete, among others.
  • Features innovative PickPen® immunomagnetic separation (IMS) for additional specificity and faster time to results
  • Simplified sample preparation, does not require heating blocks or preparation of reagents

Components

Each 576ATM (for PPMX Automation) kit includes:
  • Salmonella Amplification Tubes Tq (0.2 mL)
  • Salmonella Concentration Reagent

For each 576ATM kit, the following are also necessary and should be ordered separately:
  • 61031BC Wash Solution Kit
  • 34724BC Resuspension Buffer Tq

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

AOAC is a registered trademark of AOAC International
Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service