Skip to Content
Merck
All Photos(3)

Key Documents

71010ATMBC

Millipore

Assurance® GDS Listeria monocytogenes Tq Kit for Automation

suitable for PCR testing in Food and Beverages applications

Synonym(s):

Nucleic acid amplification system

Sign Into View Organizational & Contract Pricing

Select a Size

576 TESTS
$10,550.00

$10,550.00


Check Cart for Availability

Request more information

Select a Size

Change View
576 TESTS
$10,550.00

About This Item

UNSPSC Code:
41106302
NACRES:
NA.85

$10,550.00


Check Cart for Availability

Request more information

Product Name

GDS Listeria monocytogenes Tq Automation Kit, BioControl, Molecular based PCR test for the detection of Listeria monocytogenes in food and environmental samples

Agency

PCR, Immunomagnetic Separation (IMS)
AOAC® (PTM)
Health Canada
MicroVal

Quality Level

product line

Assurance GDS®

shelf life

6‑12 mo.

packaging

pkg of 576 tests

manufacturer/tradename

BioControl Systems 710010-576ATM

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl GDS
for use with GDS Rotor-Gene

storage temp.

2-8°C

suitability

Listeria monocytogenes

General description

GDS Listeria monocytogenes Tq Automation Kit is an automated nucleic acid amplification system for the detection of Listeria monocytogenes in food and environmental samples. The proprietary PickPen® IMS-based sample preparation procedure quickly and effectively captures Listeria monocytogenes, separating the pathogen from inhibitory background materials and ensuring valid PCR reactions for all sample types, including challenging food and environmental samples. Assurance® GDS PCR technology provides two additional levels of specificity by using both highly specific primers and probes to ensure accurate detection of Listeria monocytogenes.

Application

GDS for Listeria monocytogenes Tq is an automated nucleic acid amplification system for the detection of Listeria monocytogenes in foods, ingredients, and environmental samples

Features and Benefits

  • Results in 24 hours for environmental samples and select foods.
  • Single-step enrichment reduces handling time.
  • Validated for liquid milk, Mexican soft cheese, frankfurters, deli turkey, raw fish, raw green beans, stainless steel, rubber, concrete, plastic, among others.
  • Features innovative PickPen® immunomagnetic separation (IMS) for additional specificity and faster time to results.
  • Simplified sample prep does not require heating blocks or preparation of reagents

Components

Each 576ATM (for PPMX Automation) kit includes:
  • Listeria mono Amplification Tubes Tq (0.2 mL)
  • Listeria Concentration Reagent

For each 576ATM kit, the following are also necessary and should be ordered separately:
  • 61031BC Wash Solution Kit
  • 34745BC Listeria Resuspension Buffer Tq

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

AOAC is a registered trademark of AOAC International
Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service