Skip to Content
Merck
All Photos(1)

Key Documents

68082

Supelco

2,4′-Dibromoacetophenone

for HPLC derivatization, LiChropur, ≥99.0% (HPLC)

Synonym(s):

4′-Bromophenacyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$128.00

$128.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$128.00

About This Item

Linear Formula:
BrC6H4COCH2Br
CAS Number:
Molecular Weight:
277.94
Beilstein:
607604
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

$128.00


Check Cart for Availability

Request a Bulk Order

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

Assay

≥99.0% (HPLC)

form

solid

quality

LiChropur

technique(s)

HPLC: suitable

mp

108-110 °C (lit.)
108-112 °C

SMILES string

BrCC(=O)c1ccc(Br)cc1

InChI

1S/C8H6Br2O/c9-5-8(11)6-1-3-7(10)4-2-6/h1-4H,5H2

InChI key

FKJSFKCZZIXQIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Derivatisation reagent for HPLC separation of free fatty acids (FFAs) in human plasma[1], for liquid chromatography-UV-tandem mass spectrometric analysis of perfluorinated carboxylic acids[2].

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jinxue Qiu et al.
Journal of chromatography. A, 1235, 132-140 (2012-03-10)
The presence of perfluorocarboxylates (PFCAs) in the environment is of increasing concern due to their possible toxicity to humans and bioaccumulation in organisms. PFCAs are frequently found in river water, sediment and organisms and sometimes even in groundwater. In order
A Mehta et al.
Journal of chromatography. B, Biomedical sciences and applications, 719(1-2), 9-23 (1998-12-30)
We report a rapid and sensitive method for separation and quantitation of free fatty acids (FFAs) in human plasma using high-performance liquid chromatography (HPLC). Two established techniques of lipid extraction were investigated and modified to achieve maximal FFA recovery in
Angelo J Magro et al.
Acta crystallographica. Section D, Biological crystallography, 61(Pt 12), 1670-1677 (2005-11-23)
The crystal structure of an acidic phospholipase A(2) isolated from Bothrops jararacussu venom (BthA-I) chemically modified with p-bromophenacyl bromide (BPB) has been determined at 1.85 Angstroms resolution. The catalytic, platelet-aggregation inhibition, anticoagulant and hypotensive activities of BthA-I are abolished by
Wen-Hsin Liu et al.
Toxicology letters, 185(2), 102-109 (2009-01-03)
In view of the controversial role of catalytic activity on the cytotoxicity of phospholipase A(2) (PLA(2)), the present study is conducted to explore whether PLA(2) induces apoptotic process of human leukemia U937 cells through catalytic activity-independent pathway. Modification of His-48
Rodrigo G Stábeli et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 142(3-4), 371-381 (2006-01-31)
MjTX-II, a myotoxic phospholipase A(2) (PLA(2)) homologue from Bothrops moojeni venom, was functionally and structurally characterized. The MjTX-II characterization included: (i) functional characterization (antitumoral, antimicrobial and antiparasitic effects); (ii) effects of structural modifications by 4-bromophenacyl bromide (BPB), cyanogen bromide (CNBr)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service