Skip to Content
Merck
All Photos(1)

Documents

51833

Supelco

Sodium 1-heptanesulfonate monohydrate

≥99.0% (T)

Synonym(s):

1-Heptanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH2SO3Na · H2O
CAS Number:
Molecular Weight:
220.26
Beilstein:
3731762
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0% (T)

form

solid

mol wt

220.26 g/mol

technique(s)

ion pair chromatography: suitable

SMILES string

O.[Na+].CCCCCCCS([O-])(=O)=O

InChI

1S/C7H16O3S.Na.H2O/c1-2-3-4-5-6-7-11(8,9)10;;/h2-7H2,1H3,(H,8,9,10);;1H2/q;+1;/p-1

InChI key

XWZCREJRXRKIRQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sodium 1-heptanesulfonate monohydrate can be used as an ion-pairing (IP) reagent, to develop ion-pair reversed phase liquid chromatography method, for the determination of antituberculosis drug ethambutol hydrochloride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marisa Freitas et al.
Phytochemistry, 110, 91-103 (2015-01-03)
The intensification of agricultural productivity is an important challenge worldwide. However, environmental stressors can provide challenges to this intensification. The progressive occurrence of the cyanotoxins cylindrospermopsin (CYN) and microcystin-LR (MC-LR) as a potential consequence of eutrophication and climate change is
Determination of ethambutol by ion-pair reversed phase liquid chromatography with UV detection.
Analytica Chimica Acta, 456.2, 189-192 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service