Skip to Content
Merck
All Photos(1)

Key Documents

67779

Sigma-Aldrich

Methanethiol

purum

Synonym(s):

Mercaptan C1, Methyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SH
CAS Number:
Molecular Weight:
48.11
Beilstein:
1696840
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

1.66 (vs air)

vapor pressure

1536 mmHg ( 20 °C)

grade

purum

Assay

≥99.0%

expl. lim.

21.8 %

bp

6 °C (lit.)

mp

−123 °C (lit.)

storage temp.

2-8°C

SMILES string

CS

InChI

1S/CH4S/c1-2/h2H,1H3

InChI key

LSDPWZHWYPCBBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin with 100 mL (net ~87 g)

Other Notes

Sales restrictions may apply

control valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rima Kassim et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 18(3), 333-342 (2013-01-22)
Evaluating the reactivity of the metal-thiolate clusters in metallothionein (MT) is a key step in understanding the biological functions of this protein. The effects of the metal clustering and protein environment on the thiolate reactivity with hydrogen peroxide (H(2)O(2)) were
C Hort et al.
Waste management (New York, N.Y.), 33(1), 18-25 (2012-10-06)
This work presents the use of two composts as filter media for the treatment by biofiltration of odors emitted during the aerobic composting of a mixture containing sewage sludge and yard waste. The chemical analysis of the waste gas showed
Ivan Spasojević et al.
Amino acids, 42(6), 2439-2445 (2011-08-11)
The mechanisms of reaction of methionine with hydroxyl radical are not fully understood. Here, we unequivocally show using electron paramagnetic resonance spin-trapping spectroscopy and GC-FID and GC-MS, the presence of specific carbon-, nitrogen- and sulfur-centered radicals as intermediates of this
Michael Hulko et al.
Sensors (Basel, Switzerland), 11(6), 5968-5980 (2011-12-14)
This work is about gas biosensing with a cytochrome c biosensor. Emphasis is put on the analysis of the sensing process and a mathematical model to make predictions about the biosensor response. Reliable predictions about biosensor responses can provide valuable
Toru Takeshita et al.
Scientific reports, 2, 215-215 (2012-02-23)
Both hydrogen sulfide (H2S) and methyl mercaptan (CH(3)SH) are frequently detected in large amounts in malodorous mouth air. We investigated the bacterial composition of saliva of 30 subjects with severe oral malodor exhibiting extreme CH(3)SH/H(2)S ratios (high H(2)S but low

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service