Merck
  • The reaction of methionine with hydroxyl radical: reactive intermediates and methanethiol production.
The reaction of methionine with hydroxyl radical: reactive intermediates and methanethiol production.
Amino acids (2011-08-11)
Ivan Spasojević, Jelena Bogdanović Pristov, Ljubodrag Vujisić, Mihajlo Spasić
ABSTRACT
The mechanisms of reaction of methionine with hydroxyl radical are not fully understood. Here, we unequivocally show using electron paramagnetic resonance spin-trapping spectroscopy and GC-FID and GC-MS, the presence of specific carbon-, nitrogen- and sulfur-centered radicals as intermediates of this reaction, as well as the liberation of methanethiol as a gaseous end product. Taking into account the many roles that methionine has in eco- and biosystems, our results may elucidate redox chemistry of this amino acid and processes that methionine is involved in.
Sigma-Aldrich
Sodium thiomethoxide, 95%
Sigma-Aldrich
Sodium methanethiolate, technical, ≥90% (RT)
Sigma-Aldrich
Methanethiol, purum