Skip to Content
Merck
All Photos(1)

Key Documents

1.06683

Supelco

Potassium carbonate/sodium carbonate

GR for analysis

Synonym(s):

Potassium sodium carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$173.00
2.5 KG
$622.00

$173.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
500 G
$173.00
2.5 KG
$622.00

About This Item

UNSPSC Code:
12352402
NACRES:
NB.21

$173.00


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

form

solid

pH

11.5 (20 °C, 50 g/L in H2O)

density

2.47 g/cm3 at 20 °C

bulk density

1750 kg/m3

storage temp.

2-30°C

Application


  • Catalytic pyrolysis and in situ carbonization of walnut shells: This study highlights the use of potassium carbonate and sodium carbonate in the catalytic pyrolysis of walnut shells, demonstrating their potential to enhance the electrochemical performance of derived carbons. This process is pivotal in the development of energy storage materials and aligns with environmental sustainability goals (He W et al., 2024).

  • Determination of barium sulfate in barite ore: This research utilizes potassium and sodium carbonate in a phase conversion-partial pressure-corrected headspace gas chromatography technique for the analysis of barite ore. This method underscores the importance of carbonate salts in mineral testing and environmental analysis, providing a reliable approach for the mining and environmental sectors (Fan ZY et al., 2022).

  • Passive Fire Protection of Taeda pine Wood: This study examines the application of potassium carbonate and sodium carbonate in starch-based surface coatings for enhancing the fire resistance of Taeda pine wood. This application is critical in building material science and fire safety, showcasing the adaptability of carbonate salts in developing fire-retardant treatments (Tretsiakova-McNally S et al., 2021).

Analysis Note

Chloride (Cl): ≤ 0.002 %
Phosphate and Silicate (as SiO₂): ≤ 0.005 %
Total sulfur (as SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.002 %
Al (Aluminium): ≤ 0.001 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.001 %
Loss on ignition (600 °C): ≤ 1.0 %

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. PURITY ?

    1 answer
    1. This product is a mixture of potassium and sodium carbonate. The purity is not determined for multi-component items. Elemental analysis is reported in the product Certificate of Analysis. Please see the link below to review a sample or lot-specific Certificate:
      https://www.sigmaaldrich.com/US/en/product/mm/106683#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service