Skip to Content
Merck
All Photos(1)

Key Documents

59581

Supelco

Decanal

analytical standard

Synonym(s):

Caprinaldehyde, Decyl aldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$99.30

$99.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$99.30

About This Item

Linear Formula:
CH3(CH2)8CHO
CAS Number:
Molecular Weight:
156.27
Beilstein:
1362530
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$99.30


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

vapor pressure

~0.15 mmHg ( 20 °C)

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.428 (lit.)
n20/D 1.429

bp

207-209 °C (lit.)

density

0.83 g/mL at 20 °C
0.83 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)CCCCCCCCC

InChI

1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h10H,2-9H2,1H3

InChI key

KSMVZQYAVGTKIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Constance Oben Ayuk Enow et al.
BMC microbiology, 14, 216-216 (2014-09-03)
Analysis of the Escherichia coli collection of reference strains (ECOR) for the presence of the gene locus clyA, which encodes the pore-forming protein ClyA (cytolysin A), revealed that a non-functional clyA locus is common among certain extraintestinal pathogenic E. coli
Johanna Raffetseder et al.
PloS one, 9(11), e112426-e112426 (2014-11-12)
The standard treatment of tuberculosis (TB) takes six to nine months to complete and this lengthy therapy contributes to the emergence of drug-resistant TB. TB is caused by Mycobacterium tuberculosis (Mtb) and the ability of this bacterium to switch to
Jairo Torres Magalhães-Junior et al.
Analytical and bioanalytical chemistry, 406(26), 6691-6700 (2014-08-31)
Visceral leishmaniasis (VL) is a zoonosis whose etiologic agent in the Americas is Leishmania infantum, and dogs are the main host. Research and innovation in diagnostic techniques are essential to improve the surveillance and control of VL in endemic areas.
Nadir Abuzeid et al.
Journal of ethnopharmacology, 157, 134-139 (2014-09-28)
The emergence of multidrug-resistant strains of Mycobacterium tuberculosis underscores the need for continuous development of new and efficient methods to determine the susceptibility of isolates of Mycobacterium tuberculosis in the search for novel antimycobacterial agents. Natural products constitute an important
Che-Hong Chen et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(10), 3074-3079 (2015-02-26)
Correcting a genetic mutation that leads to a loss of function has been a challenge. One such mutation is in aldehyde dehydrogenase 2 (ALDH2), denoted ALDH2*2. This mutation is present in ∼ 0.6 billion East Asians and results in accumulation

Protocols

GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service