Merck
  • Antimycobacterial activity of selected medicinal plants traditionally used in Sudan to treat infectious diseases.
Antimycobacterial activity of selected medicinal plants traditionally used in Sudan to treat infectious diseases.
Journal of ethnopharmacology (2014-09-28)
Nadir Abuzeid, Sadaf Kalsum, Richin John Koshy, Marie Larsson, Mikaela Glader, Henrik Andersson, Johanna Raffetseder, Elsje Pienaar, Daniel Eklund, Muddathir S Alhassan, Haidar A AlGadir, Waleed S Koko, Thomas Schön, M Ahmed Mesaik, Omer M Abdalla, Asaad Khalid, Maria Lerm
ABSTRACT
The emergence of multidrug-resistant strains of Mycobacterium tuberculosis underscores the need for continuous development of new and efficient methods to determine the susceptibility of isolates of Mycobacterium tuberculosis in the search for novel antimycobacterial agents. Natural products constitute an important source of new drugs, and design and implementation of antimycobacterial susceptibility testing methods are necessary to evaluate the different extracts and compounds. In this study we have explored the antimycobacterial properties of 50 ethanolic extracts from different parts of 46 selected medicinal plants traditionally used in Sudan to treat infectious diseases. Plants were harvested and ethanolic extracts were prepared. For selected extracts, fractionation with hydrophilic and hydrophobic solvents was undertaken. A luminometry-based assay was used for determination of mycobacterial growth in broth cultures and inside primary human macrophages in the presence or absence of plant extracts and fractions of extracts. Cytotoxicity was also assessed for active fractions of plant extracts. Of the tested extracts, three exhibited a significant inhibitory effect on an avirulent strain of Mycobacterium tubercluosis (H37Ra) at the initial screening doses (125 and 6.25µg/ml). These were bark and leaf extracts of Khaya senegalensis and the leaf extract of Rosmarinus officinalis L. Further fractions of these plant extracts were prepared with n-hexane, chloroform, ethyl acetate, n-butanol, ethanol and water, and the activity of these extracts was retained in hydrophobic fractions. Cytotoxicity assays revealed that the chloroform fraction of Khaya senegalensis bark was non-toxic to human monocyte-derived macrophages and other cell types at the concentrations used and hence, further analysis, including assessment of IC50 and intracellular activity was done with this fraction. These results encourage further investigations to identify the active compound(s) within the chloroform fraction of Khaya senegalensis bark.
Sigma-Aldrich
Decanal, natural, ≥97%, FG
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Decanal, FCC, FG
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Decanal, ≥98% (GC), liquid
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Supelco
Decanal, analytical standard
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, Vetec, reagent grade, ≥99.5% (HPLC)