55420
CLED Agar
suitable for microbiology, NutriSelect® Plus
Synonym(s):
BROLACIN Agar, Bromothymol-blue Lactose Cystine Agar, Cystine-Lactose-Electrolyte Deficient Agar
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 15 g/L
beef extract, 3 g/L
bromo thymol blue, 0.02 g/L
casein enzymic hydrolysate, 4 g/L
L-cystine, 0.128 g/L
lactose, 10 g/L
peptic digest of animal tissue, 4 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbe id | utilization test: suitable
microbiological culture: suitable
final pH
7.3±0.2 (25 °C)
application(s)
clinical testing
environmental
food and beverages
microbiology
suitability
nonselective and differential for Enterococcus spp.
nonselective and differential for Escherichia coli
nonselective and differential for Klebsiella spp.
nonselective and differential for Lactobacillus spp.
nonselective and differential for Proteus spp.
nonselective and differential for Pseudomonas spp.
nonselective and differential for Salmonella spp.
nonselective and differential for Staphylococcus spp.
nonselective and differential for Streptococcus spp.
nonselective and differential for coliforms
General description
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.
An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.
Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques
Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service