Skip to Content
Merck
All Photos(1)

Key Documents

551333

Supelco

Tetramethylsilane solution

NMR reference standard, 12% in chloroform, NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$397.00

$397.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$397.00

About This Item

Empirical Formula (Hill Notation):
C4H12Si
CAS Number:
Molecular Weight:
88.22
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

$397.00


Check Cart for Availability

Request a Bulk Order

grade

NMR reference standard
analytical standard

Quality Level

concentration

12% in chloroform

technique(s)

NMR: suitable
qNMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

SMILES string

C[Si](C)(C)C

InChI

1S/C4H12Si/c1-5(2,3)4/h1-4H3

InChI key

CZDYPVPMEAXLPK-UHFFFAOYSA-N

Features and Benefits

Lineshape

Quantity

5 mm O.D. tube contains 0.700 mL.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service