Skip to Content
Merck
All Photos(1)

Key Documents

1.08183

Sigma-Aldrich

Tetramethylsilane

for the calibration of NMR spectra for NMR spectroscopy MagniSolv

Synonym(s):

Tetramethylsilane, TMS

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$250.00

$250.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$250.00

About This Item

Linear Formula:
(CH3)4Si
CAS Number:
Molecular Weight:
88.22
MDL number:
UNSPSC Code:
12352103
EC Index Number:
200-899-1
NACRES:
NA.22
grade:
NMR grade
technique(s):
NMR: suitable
bp:
26 °C/1013 hPa
vapor pressure:
750 hPa ( 20 °C)

$250.00


Check Cart for Availability

Request a Bulk Order

grade

NMR grade

Quality Level

vapor pressure

750 hPa ( 20 °C)

Assay

≥99.7% (GC)

form

liquid

autoignition temp.

345 °C (DIN 51794)

expl. lim.

1.0-37.9 % (v/v)

technique(s)

NMR: suitable

bp

26 °C/1013 hPa

mp

-95 °C

transition temp

flash point -20 °C (DIN 51755 Part 1)

solubility

0.02 g/L

density

0.65 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

[Si](C)(C)(C)C

InChI

1S/C4H12Si/c1-5(2,3)4/h1-4H3

InChI key

CZDYPVPMEAXLPK-UHFFFAOYSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Analysis Note

Purity (GC): ≥ 99.7
Identity (IR-spectrum): conforms
Density (d 20 °C/ 4 °C): 0.6385 - 0.6400
Performance test (NMR-spectrum): conforms

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service