Skip to Content
Merck
All Photos(1)

Key Documents

53442

Supelco

Hexyl-d13 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate

analytical standard

Synonym(s):

Diethylamino hydroxybenzoyl hexyl-d13 benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D13H18NO4
Molecular Weight:
410.59
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

O=C(C1=C(C(OC([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])[2H])[2H])=O)C=CC=C1)C2=C(O)C=C(N(CC)CC)C=C2

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service