Skip to Content
Merck
All Photos(1)

Key Documents

93777

Supelco

Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate

analytical standard

Synonym(s):

DHHB, Diethylamino hydroxybenzoyl hexyl benzoate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$210.00

$210.00


Available to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
$210.00

About This Item

Empirical Formula (Hill Notation):
C24H31NO4
CAS Number:
Molecular Weight:
397.51
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$210.00


Available to ship on08 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCCCOC(=O)c1ccccc1C(=O)c2ccc(cc2O)N(CC)CC

InChI

1S/C24H31NO4/c1-4-7-8-11-16-29-24(28)20-13-10-9-12-19(20)23(27)21-15-14-18(17-22(21)26)25(5-2)6-3/h9-10,12-15,17,26H,4-8,11,16H2,1-3H3

InChI key

FDATWRLUYRHCJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate is an organic compound belonging to the class of benzophenones, commercially used as UVA filter and is anticipated to provide reliable protection against free radicals and premature skin aging, mainly caused by shortwave UVA radiations.[1]

Application

Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate may be used as a test material in investigating the structural elucidation of its transformation products in chlorinated water using liquid chromatography coupled to mass spectrometry.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camila Martins Kawakami et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 99, 299-309 (2017-01-04)
The aim of the present study was to investigate the photochemical behavior of DHHB and its photostabilizing effect on avobenzone (AVO) in different sunscreen formulations. The formulations were subjected to photostability studies by HPLC and spectrophotometry. In vitro phototoxicity was
Massimiliano Gasparrini et al.
International journal of molecular sciences, 16(8), 17870-17884 (2015-08-08)
Strawberry polyphenols have been extensively studied over the last two decades for their beneficial properties. Recently, their possible use in ameliorating skin conditions has also been proposed; however, their role in preventing UVA-induced damage in cosmetic formulation has not yet
LC/MS study of the UV filter hexyl 2?[4?(diethylamino)?2?hydroxybenzoyl]?benzoate (DHHB) aquatic chlorination with sodium hypochlorite
Grbovic.G, et al.
Journal of Mass Spectrometry : Jms, 48(11), 1232-1240 (2013)
C Couteau et al.
International journal of pharmaceutics, 476(1-2), 160-163 (2014-10-01)
Given that previous studies have highlighted the anti-inflammatory nature, which is influenced by UV radiation, of a certain number of ultraviolet filters currently used in Europe, it seemed interesting to evaluate the persistence of this type of effect. The persistence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service