Skip to Content
Merck
All Photos(1)

Key Documents

50636

Supelco

Paraquat dichloride-(rings-d8) hydrate

PESTANAL®, analytical standard

Synonym(s):

1,1′-Dimethyl-4,4′-bipyridinium-2,2′,3,3′,5,5′,6,6′-d8 dichloride, Methyl viologen-d8 dichloride, Paraquat dichloride-d8

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$298.00

$298.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$298.00

About This Item

Empirical Formula (Hill Notation):
C12D8H6Cl2N2 · xH2O
Molecular Weight:
265.21 (anhydrous basis)
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

$298.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

C[N+](C([2H])=C1[2H])=C([2H])C([2H])=C1C2=C([2H])C([2H])=[N+](C)C([2H])=C2[2H].[Cl-].[Cl-]

InChI

1S/C12H14N2.2ClH/c1-13-7-3-11(4-8-13)12-5-9-14(2)10-6-12;;/h3-10H,1-2H3;2*1H/q+2;;/p-2/i3D,4D,5D,6D,7D,8D,9D,10D;;

InChI key

FIKAKWIAUPDISJ-FUIWBTGGSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Paraquat dichloride-d8 may be used as a reference standard for the determination of paraquat dichloride-d8 in potato, cereals, and pulses by liquid-chromatography coupled with tandem mass spectrometry (LC-MS/MS).[1]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and independent laboratory validation of a simple method for the determination of paraquat and diquat in potato, cereals and pulses.
Kolberg DIS, et al.
Analytical and Bioanalytical Chemistry, 104(8), 2465-2474 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service