Skip to Content
Merck
All Photos(5)

Key Documents

384097

Sigma-Aldrich

Ethyl viologen dibromide

99%

Synonym(s):

1,1′-Diethyl-4,4′-bipyridinium dibromide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$61.70
5 G
$258.00

$61.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$61.70
5 G
$258.00

About This Item

Empirical Formula (Hill Notation):
C14H18Br2N2
CAS Number:
Molecular Weight:
374.11
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$61.70


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

crystalline

technique(s)

titration: suitable

mp

278 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Br-].[Br-].CC[n+]1ccc(cc1)-c2cc[n+](CC)cc2

InChI

1S/C14H18N2.2BrH/c1-3-15-9-5-13(6-10-15)14-7-11-16(4-2)12-8-14;;/h5-12H,3-4H2,1-2H3;2*1H/q+2;;/p-2

InChI key

LCEBDKLPALDQPV-UHFFFAOYSA-L

General description

Ethyl viologen dibromide is known as 1,1-diethyl- 4,4-bipyridinium dibromide.

Application

Ethyl viologen dibromide has been used as an internal standard in the analysis of paraquat and diquat in the serum using high-performance liquid chromatography.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The quantitative analysis of paraquat in biological samples by liquid chromatography-electrospray ionization-mass spectrometry.
Wang Z, et al.
Journal of Analytical Toxicology, 35(1), 23-27 (2011)
Rapid analysis method for paraquat and diquat in the serum using ion-pair high-performance liquid chromatography.
Biological & Pharmaceutical Bulletin, 28(4), 725-728 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service