Skip to Content
Merck
All Photos(1)

Documents

49971

Sigma-Aldrich

D-Lactic Dehydrogenase from Lactobacillus leichmannii

suspension, yellow, ~1000 U/mL

Synonym(s):

(R)-Lactate:NAD+ oxidoreductase, D-Lactate Dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

biological source

bacterial (Lactobacillus leichmannii)

form

suspension

specific activity

≥30 U/mg protein

concentration

~1000 U/mL

impurities

L-Lactat Dehydrogenase, none detected
3.2 M Ammonium sulfate solution (pH ~6.2)

color

yellow

storage temp.

2-8°C

Application

In the food industry, the primary catalysis is coupled to conversion of NADH and H+ to NAD+ with diaphorase coupled with converting the non-fluorescent resazurin to the highly fluorescent substance resorufin to measure the content of D-lactate in food products.

Biochem/physiol Actions

D-lactic dehydrogenase catalyzes the conversion of pyruvate into D-lactate, with oxidation of NADH to NAD+. D-lactic dehydrogenase can also catalyze the reverse reaction, conversion of D-lactate into pyruvate with reduction of NAD+ to NADH.
D-lactic dehydrogenase catalyzes the conversion of pyruvate into D-lactate, with oxidation of NADH to NAD+. D-lactic dehydrogenase can also catalyze the reverse reaction, conversion of D-lactate into pyruvate with reduction of NAD+ to NADH.

Unit Definition

1 U corresponds to the amount of enzyme which will reduce 1 μmol of pyruvate to D-lactate per minute at pH 7.0 and 25 °C

Physical form

only partially soluble in water or buffer

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akos T Kovács et al.
Applied and environmental microbiology, 76(12), 4085-4088 (2010-04-20)
Bacillus coagulans has good potential as an industrial production organism for platform chemicals from renewable resources but has limited genetic tools available. Here, we present a targeted gene disruption system using the Cre-lox system, development of a LacZ reporter assay
Fei Su et al.
Journal of bacteriology, 193(17), 4563-4564 (2011-06-28)
Bacillus coagulans 2-6 is an efficient producer of lactic acid. The genome of B. coagulans 2-6 has the smallest genome among the members of the genus Bacillus known to date. The frameshift mutation at the start of the d-lactate dehydrogenase
Takenori Shibahara et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 67(Pt 11), 1425-1427 (2011-11-22)
A dye-linked D-lactate dehydrogenase from the aerobic hyperthermophilic archaeon Aeropyrum pernix was crystallized using the hanging-drop vapour-diffusion method with polyethylene glycol 8000 as the precipitant. The crystals belonged to the monoclinic space group P2(1), with unit-cell parameters a = 63.4
R Lamichhane-Khadka et al.
Journal of applied microbiology, 105(6), 1973-1981 (2009-01-06)
To identify factors associated with the Staphylococcus aureus pine-oil disinfectant-reduced-susceptibility (PD(RS)) mechanism and to describe one possible PD(RS) model. Comparative genomic sequencing (CGS) and microarray analysis were utilized to detect mutations and transcriptome alterations that occur in a S. aureus
Martin Engqvist et al.
The Journal of biological chemistry, 284(37), 25026-25037 (2009-07-10)
The Arabidopsis thaliana locus At5g06580 encodes an ortholog to Saccharomyces cerevisiae d-lactate dehydrogenase (AtD-LDH). The recombinant protein is a homodimer of 59-kDa subunits with one FAD per monomer. A substrate screen indicated that AtD-LDH catalyzes the oxidation of d- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service