Merck
  • Genome sequence of the thermophilic strain Bacillus coagulans 2-6, an efficient producer of high-optical-purity L-lactic acid.
Genome sequence of the thermophilic strain Bacillus coagulans 2-6, an efficient producer of high-optical-purity L-lactic acid.
Journal of bacteriology (2011-06-28)
Fei Su, Bo Yu, Jibin Sun, Hong-Yu Ou, Bo Zhao, Limin Wang, Jiayang Qin, Hongzhi Tang, Fei Tao, Michael Jarek, Maren Scharfe, Cuiqing Ma, Yanhe Ma, Ping Xu
ABSTRACT
Bacillus coagulans 2-6 is an efficient producer of lactic acid. The genome of B. coagulans 2-6 has the smallest genome among the members of the genus Bacillus known to date. The frameshift mutation at the start of the d-lactate dehydrogenase sequence might be responsible for the production of high-optical-purity l-lactic acid.
Sigma-Aldrich
D-Lactic Dehydrogenase from Staphylococcus epidermidis, lyophilized powder, ≥80 units/mg solid
Sigma-Aldrich
D-Lactic Dehydrogenase from Lactobacillus leichmannii, ammonium sulfate suspension, ≥250 units/mg protein (biuret)
Sigma-Aldrich
D-Lactic Dehydrogenase from Lactobacillus leichmannii, lyophilized powder, 150-500 units/mg protein