Skip to Content
Merck
All Photos(1)

Key Documents

46689

Supelco

Propionylpromazine hydrochloride

VETRANAL®, analytical standard

Synonym(s):

Propiopromazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2OS · HCl
CAS Number:
Molecular Weight:
376.94
Beilstein:
38502
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

Cl.CCC(=O)c1ccc2Sc3ccccc3N(CCCN(C)C)c2c1

InChI

1S/C20H24N2OS.ClH/c1-4-18(23)15-10-11-20-17(14-15)22(13-7-12-21(2)3)16-8-5-6-9-19(16)24-20;/h5-6,8-11,14H,4,7,12-13H2,1-3H3;1H

InChI key

ZFWVWZODBGTOIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[The opinion of O. Habler].
O Habler
Der Anaesthesist, 50(8), 613-614 (2001-09-15)
[Acute normovolemic hemodilution (ANH). Der Anaesthesist (2000) 49: 939-948].
M Alef
Der Anaesthesist, 50(8), 612-613 (2001-09-15)
[Effects and side effects of some combinations of often used tranquillizers for sedation in horses].
W Rohr et al.
DTW. Deutsche tierarztliche Wochenschrift, 89(7), 262-267 (1982-07-06)
R Langer et al.
Zentralblatt fur Veterinarmedizin. Reihe A, 41(7), 509-522 (1994-09-01)
The effect of Combolen (3.9 mg/kg bw s.c.) on the distribution of erythrocytes in dogs was investigated. After application of Combolen, the hematocrit of the animals decreased exponentially within 1 h by 21%. The reversal of the reduction began from
Residues of some veterinary drugs in animals and foods. Monographs prepared by the thirty-eighth meeting of the Joint FAO/WHO Expert Committee on Food Additives.
FAO food and nutrition paper, 41(4), 1-136 (1991-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service