Skip to Content
Merck
All Photos(2)

Key Documents

46443

Supelco

Spiroxamine

PESTANAL®, analytical standard

Synonym(s):

N-Ethyl-N-propyl-8-tert-butyl-1,4-dioxaspiro[4.5]dec-2-ylmethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$146.00

$146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$146.00

About This Item

Empirical Formula (Hill Notation):
C18H35NO2
CAS Number:
Molecular Weight:
297.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$146.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

description

mixture of diastereomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCN(CC)CC1COC2(CCC(CC2)C(C)(C)C)O1

InChI

1S/C18H35NO2/c1-6-12-19(7-2)13-16-14-20-18(21-16)10-8-15(9-11-18)17(3,4)5/h15-16H,6-14H2,1-5H3

InChI key

PUYXTUJWRLOUCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Spiroxamine belongs to the class of spiroketalamines. It is widely used as a broad-spectrum fungicide in cereals and grapes with selective, protective, curative and eradicative properties against powdery mildew.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Spiroxamine may be used as a reference standard for the determination of spiroxamine in honeybees during the honeybee poisoning incidents using gas chromatography coupled with tandem mass spectrometry (GC–MS/MS).[2]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Eyes

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

296.6 °F

Flash Point(C)

147 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrolysis study and extraction of spiroxamine from soils of different physico-chemical properties.
Rosales-Conrado N.
Chemosphere, 77(6), 821-828 (2009)
Frédéric Suffert et al.
Microbial ecology, 77(1), 110-123 (2018-06-08)
This study provides empirical evidence for antagonistic density dependence mechanisms driving sexual reproduction in the wheat fungal pathogen Zymoseptoria tritici. Biparental crosses with 12 increasing inoculum concentrations, in controlled conditions, showed that sexual reproduction in Z. tritici was impacted by
Małgorzata Baćmaga et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 54(4), 252-262 (2019-01-11)
Excess fungicides can pose a serious threat to the soil environment. Fungicides can lower the microbiological and biochemical activity of soil and lead to yield declines. Soils contaminated with fungicides have to be remediated to maintain the optimal function of
Kang Yu et al.
Frontiers in plant science, 9, 1195-1195 (2018-09-04)
Producing quantitative and reliable measures of crop disease is essential for resistance breeding, but is challenging and time consuming using traditional phenotyping methods. Hyperspectral remote sensing has shown potential for the detection of plant diseases, but its utility for phenotyping
Dganit Sadeh et al.
Journal of economic entomology, 112(2), 616-624 (2018-12-12)
The sweet potato whitefly Bemisia tabaci Genn (Hemiptera: Aleyrodidae) has been recorded to differentially prefer rosemary (Rosmarinus officinalis) varieties in commercial fields in Israel. As chemical signaling is a significant component in plant-insect interaction, the present study examined the involvement

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service