Skip to Content
Merck
All Photos(1)

Key Documents

46430

Sigma-Aldrich

2′-Fluoroacetophenone

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COCH3
CAS Number:
Molecular Weight:
138.14
Beilstein:
2041344
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.507 (lit.)
n20/D 1.509

bp

187-189 °C (lit.)

density

1.137 g/mL at 20 °C (lit.)

SMILES string

CC(=O)c1ccccc1F

InChI

1S/C8H7FO/c1-6(10)7-4-2-3-5-8(7)9/h2-5H,1H3

InChI key

QMATYTFXDIWACW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2′-Fluoroacetophenone may be used as starting reagent in the synthesis of ascididemin.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The enantioselective reduction of 2'-fluoroacetophenone utilizing a simplified CBS-reduction procedure.
Garrett CE, et al.
Tetrahedron Asymmetry, 13(13), 1347-1349 (2002)
Ida Nymann Petersen et al.
Chemical communications (Cambridge, England), 48(72), 9092-9094 (2012-08-07)
A new and convergent synthesis of ascididemin is presented. Using an anionic cascade ring closure as the key step, this natural product is obtained in 45% overall yield in just 6 steps starting from 2'-fluoroacetophenone. This new approach was extended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service