Skip to Content
Merck
All Photos(1)

Documents

219347

Sigma-Aldrich

3′-Fluoroacetophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COCH3
CAS Number:
Molecular Weight:
138.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.509 (lit.)

bp

81 °C/9 mmHg (lit.)

density

1.126 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1cccc(F)c1

InChI

1S/C8H7FO/c1-6(10)7-3-2-4-8(9)5-7/h2-5H,1H3

InChI key

HCEKGPAHZCYRBZ-UHFFFAOYSA-N

General description

Bioreduction of 3′-fluoroacetophenone by cells of Aspergillus terreus in phosphate buffer solution-glycerol has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving the enantioselective bioreduction of aromatic ketones mediated by Aspergillus terreus and Rhizopus oryzae: the role of glycerol as a co-solvent.
Andrade LH, et al.
Tetrahedron Asymmetry, 20(13), 1521-1525 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service