Skip to Content
Merck
All Photos(1)

Key Documents

45625

Supelco

Pindone

PESTANAL®, analytical standard

Synonym(s):

2-Pivaloyl-1,3-indandione, 1,3-Dioxo-2-pivaloylindan, 2-Trimethylacetyl-1,3-indandione, Pindone

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$119.00

$119.00


2 left in stockDetails

Request a Bulk Order

Select a Size

Change View
250 MG
$119.00

About This Item

Empirical Formula (Hill Notation):
C14H14O3
CAS Number:
Molecular Weight:
230.26
Beilstein:
2051258
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$119.00


2 left in stockDetails

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(C)C(=O)C1C(=O)c2ccccc2C1=O

InChI

1S/C14H14O3/c1-14(2,3)13(17)10-11(15)8-6-4-5-7-9(8)12(10)16/h4-7,10H,1-3H3

InChI key

RZKYEQDPDZUERB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Pindone is a first generation anticoagulant rodenticide,[1] belonging to the class of indandione rodenticide, commonly used for commensal rodent control.[2]

Application

Pindone may be used as an analytical reference standard for the quantification of the analyte in biological samples[1][2] using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion chromatography tandem mass spectrometry for simultaneous confirmation and determination of indandione rodenticides in serum
Chen H-X, et al.
Biomedical Chromatography, 23(11), 1217-1226 (2009)
Vitamin K and its therapeutic importance.
M E Mount et al.
Journal of the American Veterinary Medical Association, 180(11), 1354-1356 (1982-06-01)
Analysis and confirmation of rodenticide pindone in human plasma by IC-ESI-IT-MS
Chen H-X, et al.
Chromatographia, 70(7-8), 1201-1201 (2009)
G R Martin et al.
Australian veterinary journal, 68(7), 241-243 (1991-07-01)
The toxicity of pindone, a rabbit poison, to horses, cattle, goats, chickens, dogs and cats was investigated, using extension of prothrombin time (PT) as an index of poisoning. The daily dose of pindone, administered for 5 days, ranged from 0.3
P Santoro et al.
Bollettino della Societa italiana di biologia sperimentale, 69(4), 237-241 (1993-04-01)
A commercial grade Pindone was tested for the induction of genetic damage in male germ cells of Drosophila melanogaster by adult ingestion. Adults were starved for 4 days and then fed pindone at a concentration of 1 mM in ethyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service