Skip to Content
Merck
All Photos(2)

Key Documents

36764

Supelco

Diclobutrazol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$87.30

$87.30


Estimated to ship on19 April 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$87.30

About This Item

Empirical Formula (Hill Notation):
C15H19Cl2N3O
CAS Number:
Molecular Weight:
328.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$87.30


Estimated to ship on19 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

description

mixture of stereo isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(C)C(O)C(Cc1ccc(Cl)cc1Cl)n2cncn2

InChI

1S/C15H19Cl2N3O/c1-15(2,3)14(21)13(20-9-18-8-19-20)6-10-4-5-11(16)7-12(10)17/h4-5,7-9,13-14,21H,6H2,1-3H3

InChI key

URDNHJIVMYZFRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E I Mercer et al.
Steroids, 53(3-5), 393-412 (1989-03-01)
The effect of five sterol-biosynthesis-inhibiting (SBI) fungicides, triadimefon, triarimol, diclobutrazol, tridemorph, and fenpropimorph on the germination, growth, and chloroplast pigment and sterol content of barley seedlings has been studied. Triadimefon, triarimol, and diclobutrazol at 250 microM depressed germination and growth
Giuseppa Di Bella et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 54(5), 345-356 (2019-02-19)
Contaminants belonging to various classes, as polychlorobiphenyls, polycyclic aromatic hydrocarbons, organochlorine pesticides, organophosphorous pesticides, pyrethroid insecticides, fungicides, herbicides, synergists, carbamates, acaricides and insect growth regulators were simultaneously analysed by GC-MS/MS in 118 Italian and Tunisian culinary herbs and spices. The
Marek Łukasz Roszko et al.
PloS one, 11(12), e0167487-e0167487 (2016-12-03)
Chemical plant protection is a commonly discussed factor potentially responsible for decline in pollinators and other beneficial insect populations. Various groups of chemicals including persistent organic pollutants could impact a bee colony's welfare and are reported to be present in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service