Skip to Content
Merck
All Photos(1)

Documents

45483

Supelco

Fenamiphos

PESTANAL®, analytical standard

Synonym(s):

Phenamiphos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22NO3PS
CAS Number:
Molecular Weight:
303.36
Beilstein:
4752893
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=O)(NC(C)C)Oc1ccc(SC)c(C)c1

InChI

1S/C13H22NO3PS/c1-6-16-18(15,14-10(2)3)17-12-7-8-13(19-5)11(4)9-12/h7-10H,6H2,1-5H3,(H,14,15)

InChI key

ZCJPOPBZHLUFHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Fenamiphos is a non-fumigant nematicide, which is widely used for controlling nematodes in agricultural fields.

Application

Fenamiphos may be used as a reference standard in the determination of fenamiphos in water samples using high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transformation and adsorption of fenamiphos, f. sulfoxide and f. sulfone in Molokai soil and simulated movement with irrigation.
Lee C-C, et al.
Journal of Contaminant Hydrology, 1(1-2), 211-225 (1986)
Panneerselvan Logeshwaran et al.
Chemosphere, 252, 126549-126549 (2020-04-02)
Fenamiphos is a neurotoxic organophosphorus pesticide used widely to control pests of crops. Fenamiphos and its toxic oxidation products have been detected in surface and groundwaters. A novel enzyme capable of hydrolysing P-O-C bond of fenamiphos is purified from Microbacterium
Persistence and effects of fenamiphos on native algal populations and enzymatic activities in soil.
Megharaj M, et al.
Soil Biology and Biochemistry, 31(11), 1549-1553 (1999)
John A Lewis et al.
BMC genomics, 10, 202-202 (2009-05-01)
The wide use of organophosphorus (OP) pesticides makes them an important public health concern. Persistent effects of exposure and the mechanism of neuronal degeneration are continuing issues in OP toxicology. To elucidate early steps in the mechanisms of OP toxicity
Eleni Chanika et al.
Bioresource technology, 102(3), 3184-3192 (2010-11-30)
Two bacteria identified as Pseudomonas putida and Acinetobacter rhizosphaerae able to rapidly degrade the organophosphate (OP) fenamiphos (FEN) were isolated. Denaturating gradient gel electrophoresis analysis revealed that the two isolates were dominant members of the enrichment culture. Clone libraries further

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service