Skip to Content
Merck
All Photos(2)

Key Documents

45374

Supelco

Chloranil

PESTANAL®, analytical standard

Synonym(s):

Tetrachloro-p-benzoquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl4O2
CAS Number:
Molecular Weight:
245.88
Beilstein:
393006
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O

InChI

1S/C6Cl4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11

InChI key

UGNWTBMOAKPKBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Filatov
Physical chemistry chemical physics : PCCP, 13(1), 144-148 (2010-10-30)
Magnetic properties of the TTF-CA molecular crystal below the neutral to ionic transition temperature are studied using the embedded cluster approach in combination with density functional theory. The calculated values of the Heisenberg exchange integral between the neighboring TTF and
Shunichi Fukuzumi et al.
Journal of the American Chemical Society, 133(6), 1859-1869 (2011-01-12)
The electron-transfer and hydride-transfer properties of an isolated manganese(V)−oxo complex, (TBP8Cz)Mn(V)(O) (1) (TBP8Cz = octa-tert-butylphenylcorrolazinato) were determined by spectroscopic and kinetic methods. The manganese(V)−oxo complex 1 reacts rapidly with a series of ferrocene derivatives ([Fe(C5H4Me)2], [Fe(C5HMe4)2], and ([Fe(C5Me5)2] = Fc*)
Ali A Ensafi et al.
Colloids and surfaces. B, Biointerfaces, 84(1), 148-154 (2011-01-25)
An electrochemical method is described for the voltammetric determination of isoproterenol in the presence of uric acid using p-chloranil-carbon nanotubes paste electrode. Cyclic voltammetric, chronoamperometric, and electrochemical impedance spectroscopic methods have used to investigate the suitability of p-chloranil as a
Prashant Singh et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 75(3), 983-991 (2010-01-19)
In the present work, we report the synthesis and characterization of novel charge-transfer complexes of thiazolidine-2,4-dione (TZD) with sigma acceptor (iodine) and pi acceptors (chloranil, dichlorodicyanoquinone, picric acid and duraquinone). We also evaluated their thermal and electrochemical properties and we
Joo-Young Kim et al.
Biochemical pharmacology, 79(10), 1506-1515 (2010-01-26)
BLT2, a low-affinity leukotriene B4 (LTB4) receptor, is a member of the G-protein coupled receptor (GPCR) family and is involved in the pathogenesis of inflammatory diseases such as asthma. Despite its clinical implications, however, no pharmacological inhibitors are available. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service