Skip to Content
Merck
All Photos(1)

Key Documents

8.02940

Sigma-Aldrich

2,3-Dichloro-5,6-dicyano-p-benzoquinone

for synthesis

Synonym(s):

2,3-Dichloro-5,6-dicyano-p-benzoquinone, 4,5-Dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile, DDQ

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$125.00

$125.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$125.00

About This Item

Linear Formula:
2,3-(Cl)2-1-(=O)C6-4-(=O)-5,6-(CN)2
CAS Number:
Molecular Weight:
227.00
MDL number:
UNSPSC Code:
12352117
EC Index Number:
201-542-2
NACRES:
NA.22

$125.00


Available to ship on31 March 2025Details


Request a Bulk Order

Quality Level

form

powder

potency

86 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

mp

209-214 °C

storage temp.

2-30°C

SMILES string

ClC1=C(C(=O)C(=C(C1=O)C#N)C#N)Cl

InChI

1S/C8Cl2N2O2/c9-5-6(10)8(14)4(2-12)3(1-11)7(5)13

InChI key

HZNVUJQVZSTENZ-UHFFFAOYSA-N

Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service