Skip to Content
Merck
All Photos(1)

Key Documents

45302

Supelco

Chlorflurenol-methyl

PESTANAL®, analytical standard

Synonym(s):

Methyl 2-chloro-9-hydroxyfluorene-9-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClO3
CAS Number:
Molecular Weight:
274.70
Beilstein:
2871254
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)C1(O)c2ccccc2-c3ccc(Cl)cc13

InChI

1S/C15H11ClO3/c1-19-14(17)15(18)12-5-3-2-4-10(12)11-7-6-9(16)8-13(11)15/h2-8,18H,1H3

InChI key

LINPVWIEWJTEEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Eichler et al.
Archives of environmental contamination and toxicology, 11(2), 185-193 (1982-01-01)
Tests were conducted with the synthetic growth regulator chlorflurenol-methyl to investigate its rate of degradation in soil, leaching behavior, and possible side-effects on the soil microflora and on soil physiological processes. With two sandy soils (Ct = 1.0 and 2.58%)
J Mattsson et al.
Development (Cambridge, England), 126(13), 2979-2991 (1999-06-08)
To assess the role of auxin flows in plant vascular patterning, the development of vascular systems under conditions of inhibited auxin transport was analyzed. In Arabidopsis, nearly identical responses evoked by three auxin transport inhibitor substances revealed an enormous plasticity
L E Sieburth
Plant physiology, 121(4), 1179-1190 (1999-12-14)
To investigate possible roles of polar auxin transport in vein patterning, cotyledon and leaf vein patterns were compared for plants grown in medium containing polar auxin transport inhibitors (N-1-naphthylphthalamic acid, 9-hydroxyfluorene-9-carboxylic acid, and 2,3,5-triiodobenzoic acid) and in medium containing a
U Junghans et al.
Plant biology (Stuttgart, Germany), 6(1), 22-29 (2004-04-20)
The influence of the auxin transport inhibitors naphthylphthalamic acid (NPA) and methyl-2-chloro-9-hydroxyflurene-9-carboxylate (CF), as well as the gaseous hormone ethylene on cambial differentiation of poplar was determined. NPA treatment induced clustering of vessels and increased vessel length. CF caused a
Qiumei Shi et al.
Journal of experimental botany, 57(11), 2899-2907 (2006-07-27)
The effects of different kinds of mechanical wounding on nicotine production in tobacco plants were compared, with sand or hydroponics culture under controlled conditions. Both removal of the shoot apex and damage of the youngest unfolded leaves nos 1 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service