Skip to Content
Merck
All Photos(1)

Key Documents

44277

Supelco

Ammonium nitrate solution

0.9-1.1 M, for Ca-selective electrodes

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H4N2O3
CAS Number:
Molecular Weight:
80.04
UNSPSC Code:
26111700

Quality Level

form

liquid

concentration

0.9-1.1 M

InChI

1S/NO3.H3N/c2-1(3)4;/h;1H3/q-1;/p+1

InChI key

DVARTQFDIMZBAA-UHFFFAOYSA-O

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Ox. Liq. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine A Matulis et al.
Current biology : CB, 30(2), 222-236 (2020-01-14)
In visual systems, neurons adapt both to the mean light level and to the range of light levels, or the contrast. Contrast adaptation has been studied extensively, but it remains unclear how it is distributed among neurons in connected circuits
Roman Kuprov et al.
Journal of the Air & Waste Management Association (1995), 64(8), 957-969 (2014-09-05)
Under the National Ambient Air Quality Standards (NAAQS), put in place as a result of the Clean Air Amendments of 1990, three regions in the state of Utah are in violation of the NAAQS for PM10 and PM2.5 (Salt Lake
T Stockdale
Nutrition and health, 8(4), 207-222 (1992-01-01)
Some personal farming experiences are described, and attention is drawn to four anomalies associated with the use of ammonium nitrate. These can be explained if the changes that took place in the formulation of fertilisers around 1960 have led to
Cerium(IV) ammonium nitrate as a catalyst in organic synthesis.
Vellaisamy Sridharan et al.
Chemical reviews, 110(6), 3805-3849 (2010-04-03)
Jonathan J Buonocore et al.
Environment international, 68, 200-208 (2014-04-29)
We estimated PM2.5-related public health impacts/ton emitted of primary PM2.5, SO2, and NOx for a set of power plants in the Mid-Atlantic and Lower Great Lakes regions of the United States, selected to include varying emission profiles and broad geographic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service