Skip to Content
Merck
All Photos(1)

Key Documents

1.01201

Supelco

Ammonium peroxodisulfate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Ammonium peroxodisulfate, Ammonium persulfate, Peroxidisulfuric acid diammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$84.20
1 KG
$130.00
5 KG
$453.00
12 KG
$1,490.00

$84.20


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
$84.20
1 KG
$130.00
5 KG
$453.00
12 KG
$1,490.00

About This Item

Linear Formula:
(NH4)2S2O8
CAS Number:
Molecular Weight:
228.20
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-786-5
NACRES:
NA.21
grade:
ACS reagent
form:
solid

$84.20


Estimated to ship on28 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥98.0% (iodometric)

form

solid

potency

742 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter
≤0.04 meq/g Titrable free acid

ign. residue

≤0.05% (as SO4)

pH

3.2 (20 °C, 100 g/L in H2O)

mp

120 °C (decomposition)

density

1.98 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride, chlorate (as Cl-): ≤0.001%

cation traces

Fe: ≤0.001%
Mn: ≤0.00005%
heavy metals (as Pb): ≤0.001%

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])OO[S](=O)(=O)[O-].[N+H4].[N+H4]

InChI

1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)

InChI key

ROOXNKNUYICQNP-UHFFFAOYSA-N

Related Categories

Application


  • Ammonium Peroxodisulfate in Environmental Treatment: Demonstrated effectiveness in treating old landfill leachate through a combination of peroxodisulfate-assisted electro-oxidation and electro-coagulation, showcasing its potential in complex wastewater treatment scenarios (Ma et al., 2024).

  • Controlled Drug Release Systems: Utilized in the development of novel hydrolytic degradable crosslinked polymeric networks for controlled drug release, indicating its role in advancing pharmaceutical delivery technologies (Mehmood et al., 2023).

  • Synthesis of Microgels: Employed in the synthesis and in vitro evaluation of aspartic acid-based microgels designed for sustained drug delivery, showcasing its catalytic capabilities in polymer chemistry (Suhail et al., 2021).

  • Chemical Oxidative Polymerization: Ammonium peroxodisulfate is utilized in the chemical oxidative polymerization of methylene blue, helping elucidate the reaction mechanism and aspects of chain structure, crucial for understanding and developing new polymeric materials (Mezhuev et al., 2021).


Linkage

Replaces: AX1340; AX1340-1; AX1340-20; AX1340-3

Analysis Note

Assay (iodometric): ≥ 98.0 %
Insoluble matter: ≤ 0.005 %
Titrable free acid: ≤ 0.04 meq/g
Chloride, Chlorate (as Cl): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Mn (Manganese): ≤ 0.00005 %
Residue on ignition (as SO₄): ≤ 0.05 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service