Skip to Content
Merck
All Photos(1)

Documents

43264

Millipore

Agar Test pH 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171608
NACRES:
NA.74

composition

agar-agar, 13.0 g/L
peptone from casein, tryptic, 3.45 g/L
peptone from meat, tryptic, 3.45 g/L
sodium chloride, 5.1 g/L

Quality Level

final pH

6.0±0.2 (25 °C)

storage temp.

2-30°C

Application

For the detection of antimicrobial inhibitors in meat and organ samples together with Bacillus subtilis (BGA) spore suspension and Micrococcus luteus ATCC 9341 as test organisms.

Preparation Note

Suspend 25 g/litre Test Agar pH 6.0, autoclave (15 min at 121°C), test the pH and if necessary adjust. Cool to 50-45 °C, mix in 1mL/litre Bacillus subtilis (BGA) spore suspension. After mixing the spore suspension, immediately pour the plates and place in the refrigerator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service