Skip to Content
Merck
All Photos(4)

Key Documents

1.32369

Millipore

Anaerocult A mini

For the cultivation of obligate and facultative anaerobes, suitable for microbiology

Synonym(s):

Anaerobic growth, Anaerobic incubation

Sign Into View Organizational & Contract Pricing

Select a Size

25 SET
$142.00

$142.00


In StockDetails


Request more information

Select a Size

Change View
25 SET
$142.00

About This Item

UNSPSC Code:
41104308
NACRES:
NB.14

$142.00


In StockDetails


Request more information

Quality Level

form

solid

feature

Chemically bind oxygen quickly and completely, Creates an oxygen free (anaerobic) environment and a CO2 atmosphere.

manufacturer/tradename

Anaerocult

packaging

pkg of 25 Anaerocult A mini and 25 special incubation bags

storage condition

(Store dry and tightly closed)

color

white

application(s)

microbiology

compatibility

ANAEROTEST

storage temp.

15-25°C

General description

Anaerocult A mini is a reagent mixture that can create an anaerobic environment to enable the growth of facultative and obligatory anaerobic microorganisms. The Anaerocult A mini sachet is moistened with water and placed into a special incubation bag with the Petri dishes and an Anaerocult strip to monitor the absence of oxygen. The sachet contains components that chemically bind the oxygen quickly and completely, creating an oxygen-free but CO2-containing atmosphere within the bag. The Anaerocult A mini system is used for the anaerobic incubation of one to four Petri dishes.

Application

The Anaerocult A mini system is used for the anaerobic incubation of one to four Petri dishes for the cultivation of obligate and facultative anaerobes.

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
ANAEROTEST is a trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Anaerocult® system supports reliable anaerobe cultivation for microbiological research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service