Skip to Content
Merck
All Photos(1)

Documents

41428

Millipore

Peptone N-Z-Soy® BL4

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.85

biological source

soybean

Quality Level

Assay

≥12% total nitrogen (N) basis
≥2.4% amino-nitrogen basis

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥2.4%
total nitrogen (N), ≥12%

ign. residue

≤10%

loss

≤6% loss on drying

pH

4.0-5.0 (25 °C, 2% in H2O)

application(s)

microbiology

Application

Peptone N-Z-Soy® media additives may be used to analyze the effects of vitamin B supplementation of lactic acid produced by Lactobacillus casei.

Legal Information

N-Z-Soy is a registered trademark of Kerry Group

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of B vitamin supplementation on lactic acid production by Lactobacillus casei.
Yoo I K, et al.
Journal of Fermentation and Bioengineering, 84(2), 172-175 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service