Skip to Content
Merck
All Photos(1)

Key Documents

37215

Supelco

Potassium Iodide Starch Paper

Box with 200 strips 20 x 70 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
NACRES:
NA.21

description

semi-quantitative

reaction suitability

reagent type: oxidant

application(s)

environmental
food and beverages
general analytical

detection method

colorimetric

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Uchiyama et al.
International journal of pharmaceutics, 554, 81-86 (2018-11-06)
Staphylococcus aureus, a leading cause of serious human infections in both healthcare and community settings, is increasingly difficult to control due to expanding resistance to multiple antibiotic classes. Methicillin-resistant S. aureus (MRSA) strains have disseminated on a global scale and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service