Skip to Content
Merck
All Photos(2)

Key Documents

37060

Supelco

Cycloprothrin

PESTANAL®, analytical standard

Synonym(s):

(RS)-α-Cyano-3-phenoxybenzyl (RS)-2,2-dichloro-1-(4-ethoxyphenyl)cyclopropanecarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$352.00

$352.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$352.00

About This Item

Empirical Formula (Hill Notation):
C26H21Cl2NO4
CAS Number:
Molecular Weight:
482.36
Beilstein:
14340823
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$352.00


Estimated to ship on24 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

CCOc1ccc(cc1)C2(CC2(Cl)Cl)C(=O)OC(C#N)c3cccc(Oc4ccccc4)c3

InChI

1S/C26H21Cl2NO4/c1-2-31-20-13-11-19(12-14-20)25(17-26(25,27)28)24(30)33-23(16-29)18-7-6-10-22(15-18)32-21-8-4-3-5-9-21/h3-15,23H,2,17H2,1H3

InChI key

CGTWCAVZZBLHQH-UHFFFAOYSA-N

General description

Cycloprothrin is an insecticide, used for controlling insect pests on rice plants and vegetables.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of cycloprothrin in rice plants.
Seguchi K, et al.
Journal of Pesticide Science, 16(4), 599-607 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service