Skip to Content
Merck
All Photos(1)

Key Documents

36696

Supelco

3-Chlorotoluene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4Cl
CAS Number:
Molecular Weight:
126.58
Beilstein:
1903632
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.522 (lit.)

bp

160-162 °C (lit.)

mp

−48 °C (lit.)

density

1.072 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cccc(Cl)c1

InChI

1S/C7H7Cl/c1-6-3-2-4-7(8)5-6/h2-5H,1H3

InChI key

OSOUNOBYRMOXQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Chlorotoluene is a chlorinated solvent and a major environmental pollutant, which is primarily used in textile, peroxide, pesticide, pharmaceutical and dye industries.

Application

3-Chlorotoluene may be used as analytical reference standard for the quantification of the analyte in environmental samples using various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Martí et al.
Journal of chromatography. A, 1077(1), 68-73 (2005-07-02)
A study of the occurrence of chlorinated toluenes in Llobregat river (NE Spain) has been carried out. These compounds are currently being used in local textile industries as dye carriers and have replaced the common trichlorobenzene mixtures. Closed loop stripping
Jorge Regueiro et al.
Journal of chromatography. A, 1216(14), 2816-2824 (2008-10-28)
In the present work, a simple and fast methodology has been developed for the analysis of chlorotoluenes in water samples using solid-phase microextraction (SPME) coupled to gas chromatography-tandem mass spectrometry (GC/MS/MS). A multifactorial experimental design strategy was used for studying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service