Skip to Content
Merck
All Photos(1)

Key Documents

36687

Supelco

4-Aminopyridine

PESTANAL®, analytical standard

Synonym(s):

4-Pyridinamine, 4-Pyridylamine, 4AP, Fampridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Beilstein:
105782
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

273 °C (lit.)

mp

155-158 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1ccncc1

InChI

1S/C5H6N2/c6-5-1-3-7-4-2-5/h1-4H,(H2,6,7)

InChI key

NUKYPUAOHBNCPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Aminopyridine is a small molecule potassium channel blocker[1], which stimulates the release of glutamate from nerve terminals and induces seizures.[2] It can also be used as a drug that aims to improve ambulatory impairment, a major symptom of multiple sclerosis disease.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

312.8 °F

Flash Point(C)

156 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

4-Aminopyridine ameliorates experimental autoimmune neuritis in Lewis rats
Moriguchi K, et al.
Journal of Neuroimmunology, 305, 72-74 (2017)
Berberine attenuates convulsing behavior and extracellular glutamate and aspartate changes in 4-aminopyridine treated rats
Sadeghnia RH, et al.
Iranian Journal of Basic Medical Sciences, 20, 588-588 (2017)
Massimo Avoli et al.
Neurobiology of disease, 52, 168-176 (2012-12-29)
4-Aminopyridine (4AP, 50 μM) induces interictal- and ictal-like discharges in brain slices including parahippocampal areas such as the entorhinal cortex (EC) but the relation between these two types of epileptiform activity remains undifined. Here, by employing field potential recordings in
R I Westphalen et al.
British journal of anaesthesia, 110(4), 592-599 (2012-12-06)
Presynaptic effects of general anaesthetics are not well characterized. We tested the hypothesis that isoflurane exhibits transmitter-specific effects on neurotransmitter release from neurochemically and functionally distinct isolated mammalian nerve terminals. Nerve terminals from adult male rat brain were prelabelled with
Michael T Roberts et al.
Neuron, 78(5), 923-935 (2013-06-15)
Feedforward inhibition sharpens the precision of neurons throughout ascending auditory pathways, including the binaural neurons of the medial superior olive (MSO). However, the biophysical influence of inhibition is poorly understood, particularly at higher frequencies at which the relative phase of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service