Skip to Content
Merck
All Photos(1)

Key Documents

275875

Sigma-Aldrich

4-Aminopyridine

≥99%

Synonym(s):

4-Pyridinamine, 4-Pyridylamine, 4AP, Fampridine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$55.20
5 G
$124.00

$55.20


Available to ship on22 April 2025Details

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
1 G
$55.20
5 G
$124.00

About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Beilstein:
105782
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$55.20


Available to ship on22 April 2025Details

New, lower price on this item!

Request a Bulk Order

Assay

≥99%

form

solid

bp

273 °C (lit.)

mp

155-158 °C (lit.)

solubility

DMF: soluble(lit.)
DMSO: soluble(lit.)
THF: soluble(lit.)
acetone: soluble(lit.)
acetonitrile: soluble(lit.)
ethanol: soluble(lit.)
isopropanol: soluble(lit.)
methanol: soluble(lit.)
water: soluble(lit.)

SMILES string

Nc1ccncc1

InChI

1S/C5H6N2/c6-5-1-3-7-4-2-5/h1-4H,(H2,6,7)

InChI key

NUKYPUAOHBNCPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Aminopyridine (4-AP) blocks the voltage-dependent potassium channels in other nerve membranes[1]. 4-AP is a K+ channel blocker and causes epileptiform activity in in vitro preparations and is a potent convulsant in animals and man[2].

Application

4-Aminopyridine can be used as:
  • A reactant to prepare pharmacologically important 4-amino-N-pyridin-4-yl-butyramide hydrochloride.[3]
  • A reactant to prepare cinnamic acid derivatives as potent cholinesterase inhibitors.[4]
  • A starting material to prepare aza analogs of batracylin, a topoisomerase II inhibitor.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

312.8 °F

Flash Point(C)

156 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Bostock et al.
The Journal of physiology, 313, 301-315 (1981-01-01)
1. 4-Aminopyridine (4AP) and tetraethylammonium ions (TEA), which block voltage-dependent potassium channels in other nerve membranes, have been used to study nerve conduction in fibres of normal rat spinal roots and those demyelinated with diphtheria toxin. The pharmacological actions have
S Yamaguchi et al.
Epilepsy research, 11(1), 9-16 (1992-03-01)
The K+ channel blocker 4-aminopyridine (4-AP) causes epileptiform activity in in vitro preparations and is a potent convulsant in animals and man. In mice, 4-AP produces behavioral activation, clonic limb movements and wild running, followed by tonic hindlimb extension and
Design, synthesis and evaluation of novel cinnamic acid derivatives bearing N-benzyl pyridinium moiety as multifunctional cholinesterase inhibitors for Alzheimer′s disease
Lan J-S, et al.
Journal of Enzyme Inhibition and Medicinal Chemistry, 32(1), 776-788 (2017)
4-Aminopyridine derivatives with antiamnesic activity
Andreani A, et al.
European Journal of Medicinal Chemistry, 35(1), 77-82 (2000)
Silvia Ghirga et al.
Journal of the Optical Society of America. A, Optics, image science, and vision, 37(4), 643-652 (2020-05-14)
It has been recently demonstrated that the exposure of naive neuronal cells to light-at the basis of optogenetic techniques and calcium imaging measurements-may alter neuronal firing. Indeed, understanding the effect of light on nongenetically modified neurons is crucial for a

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service