Skip to Content
Merck
All Photos(1)

Key Documents

34031BC

Millipore

GDS Resuspension Buffer Non-Tq

BioControl, for use with GDS

Synonym(s):

Buffer for PCR Sample Prep

Sign Into View Organizational & Contract Pricing

Select a Size

6 ML
$49.70

$49.70


Estimated to ship on24 April 2025


Request more information

Select a Size

Change View
6 ML
$49.70

About This Item

UNSPSC Code:
41106306
NACRES:
NA.85

$49.70


Estimated to ship on24 April 2025


Request more information

shelf life

36 mo.

Quality Level

packaging

bottle of 6 mL

manufacturer/tradename

BioControl

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS

storage temp.

2-8°C

suitability

Escherichia coli (O157:H7)
enterovirulent E. coli (EEC) (Shiga Toxigenic)

General description

For use with specific GDS Application Notes only.

Application

For use in Application Note MK_AN4580EN / MS_AN4580EN, cannot be substituted with GDS Resuspension Buffer Tq.

Components

One bottle, 70 mL

Compatibility

BioControl GDS

Testing Method

PCR

also commonly purchased with this product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service