Skip to Content
Merck
All Photos(1)

Key Documents

34746BC

Millipore

Assurance® GDS Listeria DNA Swab Resuspension Buffer Tq PCR Reagent

For use with Assurance® GDS Listeria Tq

Synonym(s):

Buffer for PCR Sample Prep

Sign Into View Organizational & Contract Pricing

Select a Size

1.2 ML
$32.30

$32.30


Check Cart for Availability

Request more information

Select a Size

Change View
1.2 ML
$32.30

About This Item

UNSPSC Code:
41106306
NACRES:
NA.85

$32.30


Check Cart for Availability

Request more information

Product Name

GDS Listeria DNA Swab Resuspension Buffer Tq, BioControl, For use with GDS Listeria

Quality Level

form

liquid

shelf life

24 mo.

packaging

vial of 1.2 mL (sufficient for 100 tests)

manufacturer/tradename

BioControl Systems 34746-100

storage condition

(Tightly closed in a dry, cool, and well-ventilated place)

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with BioControl GDS

storage temp.

2-8°C

suitability

Listeria spp.

Related Categories

General description

GDS Listeria DNA Swab Resuspension Buffer Tq is a requisite recommended for the labs to have onsite, in case of a contamination event. Improper handling and disposal of used Assurance® GDS amplification tubes may result in DNA amplicon contamination of laboratory work surfaces and equipment. As a preventive measure, it is recommended to perform periodic sampling for DNA contamination. GDS Listeria DNA Swab Resuspension Buffer Tq is transferred to the appropriate amplification tubes for all Assurance® GDS Listeria assays.

Application

GDS Listeria DNA Swab Resuspension Buffer Tq is used during environmental sampling for Listeria DNA contamination.

Components

One vial, 1.2 mL, sufficient for 100 tests

Testing Method

PCR

Other Notes

Listeria resuspension Buffer Tq from GDS kits CANNOT be substituted for Listeria DNA Swab Resuspension Buffer Tq.

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service