Skip to Content
Merck
All Photos(2)

Key Documents

33881

Supelco

AMOZ-d5

VETRANAL®, analytical standard

Synonym(s):

3-Amino-5-morpholinomethyl-2-oxazolidinone-d5, 4,4,5,α,α-Pentadeutero-3-amino-5-morpholinomethyl-2-oxazolidinone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$375.00

$375.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$375.00

About This Item

Empirical Formula (Hill Notation):
C8H10D5N3O3
CAS Number:
Molecular Weight:
206.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$375.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+5

SMILES string

[2H]C([2H])(N1CCOCC1)C2([2H])OC(=O)N(N)C2([2H])[2H]

InChI

1S/C8H15N3O3/c9-11-6-7(14-8(11)12)5-10-1-3-13-4-2-10/h7H,1-6,9H2/i5D2,6D2,7D

InChI key

TVHAMVOINIHMEX-VJPLVGRJSA-N

General description

AMOZ-d5 is a labeled deuterated metabolite of nitrofuran, an antimicrobial drug used in veterinary medicine.[1]

Application

AMOZ-d5 can be used as an internal standard for the determination of AMOZ in meat samples[1] and aquaculture products[2] using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the furaltadone metabolite 5-methylmorpholino-3-amino-2-oxazolidinone (AMOZ) using liquid chromatography coupled to electrospray tandem mass spectrometry during the nitrofuran crisis in Portugal.
Barbosa J, et al.
Accreditation and Quality Assurance, 12(10), 543-551 (2007)
Quantitation of chloramphenicol and nitrofuran metabolites in aquaculture products using microwave-assisted derivatization, automated SPE, and LC-MS/MS.
Veach TB, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 98(3), 588-594 (2015)
Wei Jia et al.
Analytica chimica acta, 957, 29-39 (2017-01-22)
An analytical method was developed and validated for simultaneous analysis of one hundred and thirty-seven veterinary drug residues and metabolites from sixteen different classes in tilapia utilizing an improved fully non-targeted way of data acquisition with fragmentation. The automated on-line
Effects of cyazofamid against Plasmodiophora brassicae Woronin on Chinese cabbage.
Mitani S, et al.
Pest Management Science, 59(3), 287-293 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service