Skip to Content
Merck
All Photos(2)

Key Documents

33868

Supelco

2-NP-AOZ

VETRANAL®, analytical standard

Synonym(s):

3-(2-Nitrobenzylidenamino)-2-oxazolidinone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$231.00

$231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$231.00

About This Item

Empirical Formula (Hill Notation):
C10H9N3O4
CAS Number:
Molecular Weight:
235.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$231.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

[O-][N+](=O)c1ccccc1\C=N\N2CCOC2=O

InChI

1S/C10H9N3O4/c14-10-12(5-6-17-10)11-7-8-3-1-2-4-9(8)13(15)16/h1-4,7H,5-6H2/b11-7+

InChI key

OHYXOGZWSSNLON-YRNVUSSQSA-N

General description

2-NP-AOZ is a 2-nitrophenyl derivative of the tissue-bound metabolite, 3-amino-2-oxazolidinone (AOZ).[1]

Application

2-NP-AOZ may be used as a reference standard for the determination of 2-NP-AOZ in aquatic animals using enzyme linked immunosorbent assay (ELISA).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Derivatized standard for the assay of nitrofurans in animal tissue by LC/MS[3]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and Residue Screening of the Furazolidone Metabolite, 3-Amino-2-oxazolidinone (AOZ), in Cultured Fish by an Enzyme-Linked Immunosorbent Assay.
Cheng C-C, et al.
Journal of Agricultural and Food Chemistry, 57(13), 5687?5692-5687?5692 (2009)
Application of a modified enzyme-linked immunosorbent assay for 3-amino-2-oxazolidinone residue in aquatic animals.
Liu Y, et al.
Analytica Chimica Acta, 664(13), 151-157 (2010)
A. Conneely et al.
Analytica Chimica Acta, 483, 91-91 (2003)
Fadi Aldeek et al.
Journal of agricultural and food chemistry, 66(20), 5018-5030 (2017-12-29)
We developed and validated a method for the extraction, identification, and quantitation of four nitrofuran metabolites, 3-amino-2-oxazolidinone (AOZ), 3-amino-5-morpholinomethyl-2-oxazolidinone (AMOZ), semicarbazide (SC), and 1-aminohydantoin (AHD), as well as chloramphenicol and florfenicol in a variety of seafood commodities. Samples were extracted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service