Skip to Content
Merck
All Photos(1)

Documents

32443

Supelco

Carboxyibuprofen

VETRANAL®, analytical standard

Synonym(s):

2-[4-(2-Carboxypropyl)phenyl]propionic acid, 3-[4-(1-Carboxyethyl)phenyl]-2-methylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16O4
CAS Number:
Molecular Weight:
236.26
Beilstein:
5951324
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(Cc1ccc(cc1)C(C)C(O)=O)C(O)=O

InChI

1S/C13H16O4/c1-8(12(14)15)7-10-3-5-11(6-4-10)9(2)13(16)17/h3-6,8-9H,7H2,1-2H3,(H,14,15)(H,16,17)

InChI key

DIVLBIVDYADZPL-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samantha A Whiteside et al.
Journal of medical microbiology, 68(8), 1244-1252 (2019-06-12)
The high incidence of urinary tract infection (UTI) among women and children, in combination with a lack of antibiotic efficacy with regard to pathogen eradication and recurrence prevention, as well as the negative side effects associated with antibiotics, has led
Emilia Waraksa et al.
Journal of separation science, 41(20), 3881-3891 (2018-08-14)
Ibuprofen is widely used in human and veterinary medicine for the treatment of chronic pain as well as rheumatic and musculoskeletal disorders. However, the analgesic and anti-inflammatory properties of Ibuprofen have contributed to frequent drug abuse in equestrian sports. A
Dolores Camacho-Muñoz et al.
Journal of mass spectrometry : JMS, 52(2), 94-108 (2017-01-07)
This paper presents a multi-residue method for direct enantioselective separation of chiral pharmacologically active compounds in environmental matrices. The method is based on chiral liquid chromatography and tandem mass spectrometry detection. Simultaneous chiral discrimination was achieved with a macrocyclic glycopeptide-based
Aleksandar Lolić et al.
The Science of the total environment, 508, 240-250 (2014-12-08)
The occurrence of seven pharmaceuticals and two metabolites belonging to non-steroidal anti-inflammatory drugs and analgesics therapeutic classes was studied in seawaters. A total of 101 samples covering fourteen beaches and five cities were evaluated in order to assess the spatial
Paula Paíga et al.
Journal of pharmaceutical and biomedical analysis, 106, 61-70 (2014-07-09)
An analytical methodology for the simultaneous determination of seven pharmaceuticals and two metabolites belonging to the non-steroidal anti-inflammatory drugs (NSAIDs) and analgesics therapeutic groups was developed based on off-line solid-phase extraction and ultra-high performance liquid chromatography coupled to tandem mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service